BioFX® Chemiluminescent Sensitive Plus HRP Microwell and/or Membrane Substrate, Surmodics IVD
Supplier: Surmodics
LERI-0110-2C
101396-952EA 200 USD
101396-952
BioFX® Chemiluminescent Sensitive Plus HRP Microwell and/or Membrane Substrate, Surmodics IVD
Electrophoresis Reagents Western Blotting Reagents
Chemiluminescent substrates provide a higher degree of sensitivity than other substrates to enable lower limits of detection to be achieved in ELISA and Western Blot assays. The increase in sensitivity requires more vigilance during the optimization process through the proper selection of the luminol substrate, antibody/antigen titers and blockers. Surmodics provides multiple 2-component HRP chemiluminescent substrates with various intensities to allow the assay developer to choose the appropropriate substrate for the application.
  • Picogram to femtogram levels of sensitivity
  • High signal-to-noise ratios
BioFX® Chemiluminescent Sensitive Plus HRP Substrate provides detection of peroxidase in ELISA and Western Blot applications.
Certifications: ISO 13485:2016 & 9001:2015 Certified