BioFX® Chemiluminescent Ultra Sensitive AP Microwell and/or Membrane Substrate (450 nm), Surmodics
Supplier: Surmodics
APU4-0100-01
101396-882EA 375 USD
101396-882
BioFX® Chemiluminescent Ultra Sensitive AP Microwell and/or Membrane Substrate (450 nm), Surmodics
Electrophoresis Reagents Western Blotting Reagents
Chemiluminescent substrates provide a higher degree of sensitivity than other substrates to enable lower limits of detection to be achieved in ELISA and membrane assays. The increase in sensitivity requires more vigilance during the optimization process through the proper selection of the dioxetane substrate, antibody/antigen titers and blockers. Surmodics provides multiple 1-component AP chemiluminescent substrates with various intensities and wavelength emissions to allow the assay developer to choose the appropropriate substrate for the application.
  • Absorbance at 450 nm
  • High signal to noise ratios
A one-component dioxetane chemiluminescent substrate that provides ultra-sensitive detection of alkaline phosphatase in ELISA and membrane applications.
Certifications: ISO 13485:2016 & 9001:2015 Certified