Glass Inserts for Vials, PerkinElmer
Supplier: PerkinElmer
N9300703
10139-786PK 207.65 USD
10139-786
Glass Inserts for Vials, PerkinElmer
Vials
Clear glass inserts for HPLC and GC autosampler vials are rimless and have optimum visiblity.
Glass inserts allow for maximum sample recovery and easier sample removal because the conical shape in most models decreases the surface area inside the vial.