ESI MP² Two-Stop ICP Peristaltic Pump Tubing, PerkinElmer
Supplier: PerkinElmer
N8145145 N8145150 N8145151 N8145153 N8145154 N8145156 N8145158 N8145159 N8145160 N8145161 N8145162 N8145148 N8145149 N8145180 N8145181 N8145183 N8145184 N8145185 N8145186 N8145187 N8145188 N8145189 N8145191 N8145206 N8145207 N8145208 N8145209 N8145210 N8145212 N8145216 N8145217 N8145218 N8145219 N8145165 N8145171 N8145173 N8145174 N8145175 N8145193 N8145194 N8145195 N8145196 N8145197 N8145198 N8145199 N8145200 N8145201 N8145202 N8145214 N8145215
10136-732PK 112.7 USD
10136-732 10136-734 10136-736 10136-778 10136-780 10136-782 10136-784 10136-786 10136-788 10136-790 10136-792 10136-794 10136-796 10136-798 10136-800 10136-802 10139-552 10139-554 10139-556 10139-558 10139-560 10139-562 10139-564 10139-566 10139-568 10139-570 10139-572 10139-574 10139-576 10139-578 10139-580 10139-582 10139-584 10139-586 10139-588 10139-590 10139-592 10139-594 10139-596 10139-598 10139-600 10139-602 10139-604 10139-606 10139-608 10139-610 10139-612 10139-678 10139-680 10139-682 10139-684 10139-686 10139-688 10139-690 10139-692 10139-694 10139-696 10139-698 10139-700 10139-702 10139-704 10139-706 10139-708 10139-710 10139-712 10139-714 10139-716 10139-718 10139-720 10139-722 10139-754 10139-756 10139-758 10139-760 10139-762 10139-764
ESI MP² Two-Stop ICP Peristaltic Pump Tubing, PerkinElmer
Tubing Transfer Tubing
Tubing for ESI MP² micro peristaltic pumps.
  • Non-flared and flared tubes available
  • Bridge length of 72 mm
Flared tubes for easy insertion of PFA capillaries.