Holmium Oxide Glass Filter, PerkinElmer
Supplier: PerkinElmer
N1010545
10139-532EA
1660.91
USD
10139-532
Holmium Oxide Glass Filter, PerkinElmer
Standards
Color Reference and Spectroscopy Standards
Holmium oxide glass filter for checking the wavelength accuracy of a UV/Vis or UV/Vis/NIR spectrometer.
- Glass construction
- Mounted on aluminum spring mount
- Provided in a robust aluminum container
The filter is mounted in precision aluminum spring mount and is provided in a robust aluminum container, with a Certificate of Calibration.