Sample Loops for Oil Samples, PerkinElmer
Supplier: PerkinElmer
N0777594
10139-522EA 373.12 USD
10139-522
Sample Loops for Oil Samples, PerkinElmer
Chromatography Supplies Chromatography Sample Loops
The PFA sample loop is used with the AXPRESS® PLUS valve/pump module for oils applications.
The assembly includes 2 flangeless connectors.