Valve for ASXPRESS PLUS, 6-Port, PerkinElmer
Supplier: PerkinElmer
N0777575
10139-508EA 3309.07 USD
10139-508
Valve for ASXPRESS PLUS, 6-Port, PerkinElmer
Valves Injection Valves
This 6-Port valve, designed for use with aqueous applications, is specifically designed for the ASXPRESS® PLUS.
  • 1.52 mm diameter ports
  • Labels included
The valve, manufactured from polyphenylene sulfide (PPS), contains six ports, each 1.52 mm in diameter. Labels for the ports are included.