Carbon Fiber Sample Probe, 10" for ASXPRESS® PLUS, PerkinElmer
Supplier: PerkinElmer
N0777568
10139-504EA 1277.18 USD
10139-504
Carbon Fiber Sample Probe, 10" for ASXPRESS® PLUS, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
Carbon fiber sample probe, with Int.Ø of 1.0 mm, for the ASXPRESS® PLUS.
The ASXPRESS® PLUS FEP-covered carbon fiber sample probe is 10" in length, and with tubing, has a capillary length of 75".
Delivery information: This probe kit includes PFA capillary tubing, capillary tube connection fitting, (2) probe clamps, ferrule, and (5) O-Rings.