Autosampler Carbon Fiber Sample Probe, PerkinElmer
Supplier: PerkinElmer
N0777547
10139-496EA 1128.1 USD
10139-496
Autosampler Carbon Fiber Sample Probe, PerkinElmer
Autosamplers
The carbon fiber sample probe is designed for use with the ASX-260, ASX-280, ASX-520, ASX-520HS, ASX-560, EXR-8, XLR-8 and XLR-860 instruments.
  • 0.5 mm Int.Ø
  • Blue band
Carbon fiber reinforced probes are designed to be mounted to a Z-drive with two probe clamps and without a guide plate. The carbon fiber probes can also be used with the prior version Z-drives, but the guide plate must be used. Most CETAC autosamplers built since January 2009 are equipped with carbon fiber probes and associated Z-drives. (Septum-piercing autosamplers are an exception.) All wetted surfaces (liquid contact) of both the carbon fiber and Ultem probes are inert, identical, and made of PTFE.