PFA FAST SC Autosampler Probe, 1.0 mm int.Ø., PerkinElmer
Supplier: PerkinElmer
N0777522
10139-488EA 1766.5 USD
10139-488
PFA FAST SC Autosampler Probe, 1.0 mm int.Ø., PerkinElmer
Autosamplers
PFA FAST probe offers high chemical resistance for applications involving strong acids, alkalis and organics.
  • Contoured design reduces carryover
  • PFA material provides a high chemical resistance
  • Carbon fiber support
  • Compatible with SC-8 and SC-14 autosamplers
A contoured design reduces carryover, while an all PFA wetted sample flow path reduces contamination. SC Autosampler Probe has a carbon fiber support, 1.0 mm I.D. (gray marker), 150 cm length, connects to port #5 (1/4-28 fitting) high flow.