SeaSpray™ Glass Nebulizer for NexION, Elan®, and Optima™ Instruments, PerkinElmer
Supplier: PerkinElmer
N0777484
10139-472EA 2613.2 USD
10139-472
SeaSpray™ Glass Nebulizer for NexION, Elan®, and Optima™ Instruments, PerkinElmer
Atomizers
The SeaSpray™ nebulizer is designed for use with ELAN® 5000, 6x00, 9000, and DRCs and NexION® 300 and 350 ICP-MS series instruments, as well as Optima® 3x00 RL ICP-OES series instruments.
  • Glass nebulizer
  • 0.4 ml/min. uptake rate
The SeaSpray™ concentric nebulizer is ideal for samples with high dissolved solids and offers outstanding nebulization efficiency for trace level analysis. It offers freedom from clogging while nebulizing solutions near the limit of solubility of most mineral salts and conferring significant sensitivity gains. They are supplied with the EzyLok Argon connector.