EzyFit Connector for SeaSpray and MicroMist Nebulizers, PerkinElmer
Supplier: PerkinElmer
N0777464
10139-440PK 2808.38 USD
10139-440
EzyFit Connector for SeaSpray and MicroMist Nebulizers, PerkinElmer
Atomizers
This EzyFit Connector is for use with MicroMist and SeaSpray nebulizers in Optima® 4x00/5x00/7x00/8x00 and Avio® 200/500 ICP-OES series instruments in addition to ELAN® 5000/6x00/9000/DRCs and NexION® 300/350/1000/2000 ICP-MS series instruments.
This product serves as a replacement connector and sample tubing for glass concentric nebulizers with EzyFit sample inlet. This sample tube is replacement tubing for internal standard kits. It is included in the HF-Resistant Internal Standard Kit (10139-142) and the Non-HF-Resistant Internal Standard Kit (10139-144).
Delivery information: Includes 10 connectors.