EzyFit Connector for SeaSpray™ and MicroMist Nebulizers , PerkinElmer
Supplier: PerkinElmer
N0777463
10139-438PK 729.66 USD
10139-438
EzyFit Connector for SeaSpray™ and MicroMist Nebulizers , PerkinElmer
Atomizers
The EzyFit Connector is designed for use with MicroMist and SeaSpray nebulizers in Optima® 4x00/5x00/7x00/8x00 and Avio® 200/500 ICP-OES series instruments in addition to ELAN® 5000/6x00/9000/DRCs and NexION® 300/350/1000/2000 ICP-MS series instruments.
  • Replacement connector and sample tubing for glass concentric nebulizers with EzyFit sample inlet
  • 700 mm length
  • Includes 10 connectors
The EzyFit Twin Sample Tube, a 0.50 mm Int.Ø. × ¹/₁₆" Ext.Ø × 700 mm long version of this EzyFit connector, can be purchased separately. This sample tube is replacement tubing for internal standard kits. It is included in the HF-resistant internal standard kit and the non HF-resistant internal standard kit.