Santoprene Two-Stop Peristaltic Pump Tubing, PerkinElmer
Supplier: PerkinElmer
N0777444
10139-410PK 221.96 USD
10139-410
Santoprene Two-Stop Peristaltic Pump Tubing, PerkinElmer
Tubing Transfer Tubing
This Santoprene™ two-stop pump tubing is for use with the standard pump in PerkinElmer Optima, Avio, and NexION 300/350 series instruments.
Packaging: This tubing ships as a package of 12.