F5-50 Solution Fitting for Standard MEINHARD™ Nebulizers, PerkinElmer
Supplier: PerkinElmer
N0777414
10139-384EA 153.77 USD
10139-384
F5-50 Solution Fitting for Standard MEINHARD™ Nebulizers, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This Standard Nebulizer Solution Fitting is designed for use with Optima™ 4x00/5x00/7x00/8x00 and Avio® 200/500 ICP-OES series instruments.
  • PTFE capillary measures 0.5 mm I.D.×70 cm in length
This nebulizer fitting is included with all standard MEINHARD™ Glass Nebulizers.