EzyLok Kit for Glass Concentric Nebulizers, PerkinElmer
Supplier: PerkinElmer
N0777413
10139-382EA 185.51 USD
10139-382
EzyLok Kit for Glass Concentric Nebulizers, PerkinElmer
Atomizers
This EzyLok kit is designed for use with Optima™ 4×00/5×00/7×00/8×00 and Avio® 200/500 ICP-OES series instruments. It is designed for use with Conikal™, slurry, SeaSpray™, and MicroMist™ nebulizers.
The EzyLok kit is included in the assembly with the Conikal™ nebulizer, 2 ml/min(10138-152), slurry nebulizer, 4 ml/min (10138-154), SeaSpray™ nebulizer, 2 ml/min(10138-174), MicroMist™ nebulizer, 0.6 ml/min (10138-176), MicroMist™ nebulizer, 0.2 ml/min (101690-960), MicroMist™ nebulizer, 0.1 ml/min (101690-962), and the MicroMist™ nebulizer, 0.05 ml/min (10138-178) for Optima 4×00/5×00/7×00/8×00 and Avio 200/500. This kit includes EzyLok connector for 4 mm tubing,  EzyLok 6 mm hose adapter, and nebulizer hose clip.