Demountable Quartz Injector, 1.0 mm Int.Ø., PerkinElmer
Supplier: PerkinElmer
N0777410
10139-376EA 2249.52 USD
10139-376
Demountable Quartz Injector, 1.0 mm Int.Ø., PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This Quartz Injector Cassette Mount is designed for use with demountable torches in NexION® 300/350 ICP-MS series instruments.
  • Demountable design allows the injector to be removed and replaced efficiently without dismounting the torch
Standard Injectors have an O-Ring free direct connection to Cyclonic and PFA spray chambers while O-Ring free Injectors have an O-Ring free seal that reduces contamination, makes it easy to clean and easy to insert. This injector can be used with the 0-Slot Demountable Quartz Torch (N8122006). A 1.0 mm I.D. O-Ring Free Injector is available separately (101686-298).