Bottle, PFA, with Cap, PerkinElmer
Supplier: PerkinElmer
N0777406 N0777407
10139-372PK 2806.42 USD
10139-372 10139-374
Bottle, PFA, with Cap, PerkinElmer
Bottles Plastic Bottles
PFA bottles, with cap, are available in multiple capacities and dimensions.
  • Multiple sizes available
  • Supplied with cap
PFA bottles are ideal for a wide range of demanding fluid handling and storage applications in general laboratory, semiconductor, and environmental industries.