Platinum Injector Shield Disc for ELAN Bayonet, PerkinElmer
Supplier: PerkinElmer
N0777366
10139-342EA 1587.93 USD
10139-342
Platinum Injector Shield Disc for ELAN Bayonet, PerkinElmer
Chromatography Supplies Chromatography Injectors
The platinum injector shield disc for ELAN Bayonet protects PFA injector base from damage caused by heat and light from the instrument, extends injector lifetime, and improves long-term instrument stability.
Complete high purity sample introduction system for NexION 300/350. All wetted surfaces are PTFE and Pt. Used by a majority of semiconductor labs worldwide for the analysis of semiconductor grade chemicals and Si wafer samples.