Polypropylene Autosampler Tubes, PerkinElmer
Supplier: PerkinElmer
N0777156 N0777159 N0777160 N0777161 N0777167 N0777168
101681-806PK 484.26 USD
101681-806 101687-200 101692-362 101676-680 10139-260 10139-266 10139-268 10139-270 10139-272 10139-274
Polypropylene Autosampler Tubes, PerkinElmer
Tubes Test Tubes
Polypropylene tubes are designed to provide excellent chemical resistance.
  • Translucent
  • Conical design
Produced for routine everyday applications and storage needs. Specialized models are made from different qualities of resin for higher durability.