PolyScience® Replacement Air Filter, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N0777095
10139-242EA 160.87 USD
10139-242
PolyScience® Replacement Air Filter, PerkinElmer
Filters Gas Filters
This replacement air filter is designed for use with the PolyScience® WhisperCool™ refrigerated chiller in ICP-MS and ICP-OES instruments. This 13×14" air filter with frame is fully washable and removes quickly for easy cleaning.
Filter is designed specifically for use with the PolyScience® WhisperCool™ refrigerated chillers.