Sample Loop for Aqueous Samples, 2.0 mm, PerkinElmer
Supplier: PerkinElmer
N0777087
10139-232EA 268.61 USD
10139-232
Sample Loop for Aqueous Samples, 2.0 mm, PerkinElmer
Chromatography Supplies Chromatography Sample Loops
The FEP sample loop, with a 2.0 mm inner diameter, is used with the AXPRESS® PLUS value/pump module for aqueous applications.
This inner volume capacity is 3.5 ml.
Delivery information: The full assembly includes two connectors and two ferrules.