UniFit Connector for SeaSpray and MicroMist Nebulizers -700 mm Long Sample Tube, PerkinElmer
Supplier: PerkinElmer
N0774080
10139-152PK 255.84 USD
10139-152
UniFit Connector for SeaSpray and MicroMist Nebulizers -700 mm Long Sample Tube, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This UniFit Connector is designed for use with concentric glass nebulizers in Optima® 4x00/5x00/7x00/8x00 and Avio® 200/500 ICP-OES series instruments in addition to ELAN® 5000/6x00/9000/DRCs and NexION® 300/350/1000/2000 ICP-MS series instruments. The UniFit connector is a convenient drain connector for glass cyclonic spray chambers and is 700 mm long.
This assembly includes 3 UniFit connectors.