UniFit Connector for SeaSpray™ and MicroMist™ Nebulizers, PerkinElmer
Supplier: PerkinElmer
N0774077
10139-150PK 255.9 USD
10139-150
UniFit Connector for SeaSpray™ and MicroMist™ Nebulizers, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This UniFit Connector is designed for use with concentric glass nebulizers in Optima™ 4x00/5x00/7x00/8x00 and Avio® 200/500 ICP-OES series instruments in addition to ELAN® 5000/6x00/9000/DRCs and NexION® 300/350/1000/2000 ICP-MS series instruments.
  • UniFit connector is a convenient drain connector for glass cyclonic spray chambers
  • 700 mm overall length
  • 0.75Int.Ø.×1.3Ext.Ø mm
This assembly includes 3 UniFit connectors.