SeaSpray™ Quartz Nebulizer for NexION®, ELAN®, and Optima® Instruments, PerkinElmer
Supplier: PerkinElmer
N0774069
10139-146EA 3866.49 USD
10139-146
SeaSpray™ Quartz Nebulizer for NexION®, ELAN®, and Optima® Instruments, PerkinElmer
Atomizers
This concentric SeaSpray™ Quartz Nebulizer is designed for use with PerkinElmer ELAN® 5000/6x00/9000/DRCs and NexION® 300/350/1000/2000 ICP-MS series of ICP-MS instruments. It offers freedom from clogging while nebulizing solutions near the limit of solubility of most mineral salts and conferring significant sensitivity gains.
  • 1.0 L/min. Argon flow rate
  • 1 mL/min. uptake rate
  • Ideal for samples with high dissolved solids
  • Outstanding nebulization efficiency for trace level analysis
All SeaSpray™ Nebulizers are supplied with the EzyLok Argon connector.