Silver Bromide Circular Demountable Cell Window, Thickness: 2 mm, One Pair, PerkinElmer
Supplier: PerkinElmer
L1272102
10139-110EA 2056.87 USD
10139-110
Silver Bromide Circular Demountable Cell Window, Thickness: 2 mm, One Pair, PerkinElmer
Specimen Mounting Supplies
Silver Bromide circular demountable cell windows are minimally degraded by water making them useful for applications in the deep infrared.  
One pair of cell windows 25 mm in diameter.