Circular Demountable Cell Windows, PerkinElmer
Supplier: PERKINELMER U.S. LLC
L1271102 L1271202 L1271602
10136-368EA 345.15 USD
10136-368 10139-106 10139-108
Circular Demountable Cell Windows, PerkinElmer
Specimen Mounting Supplies
Circular windows are used to construct a demountable cell where the sample is placed on one window and the second window is pressed lightly on top of the first.
  • Spacers of various thicknesses can be used to vary the cell pathlength
The windows are demountable and can be easily changed and repolished when necessary.
Ordering information: Spacers and cell parts should be ordered separately.