Silicone antifoam emulsion 10% active compound, Dow Corning® DB 110 A
Supplier: PerkinElmer
Synonyms: Anti-foaming agent
B0507226
10139-070EA 2714.51 USD
10139-070
Silicone antifoam emulsion 10% active compound, Dow Corning® DB 110 A
Silicone antifoam
Controls foaming during reactions when using the MHS-10 or MHS-20 and transits that can affect the analytical results.
  • Silicone-based compound
  • Emulsion spreads silicone fast and effectively
  • Heavy-duty defoamer
  • Suitable in non-aqueous foaming situations
Silicone-based defoamers might be delivered as an oil or a water-based emulsion, and can be used in non-aqueous foaming systems like crude oil and oil refining.