Organics High Sensitivity Nebulizer Complete Assembly with Tantalum Needle for AAnalyst, PerkinElmer
Supplier: PERKINELMER U.S. LLC
B0505590
10139-052EA 5644.14 USD
10139-052
Organics High Sensitivity Nebulizer Complete Assembly with Tantalum Needle for AAnalyst, PerkinElmer
Atomizers
This high sensitivity nebulizer assembly, with Tantalum needle and Kalrez O-rings, is for use with AAnalyst® 100/300/200/400/700/800 and earlier AA series instruments. This complete assembly with Tantalum capillary assembly is for use with solutions containing aqua regia. It provides maximum sensitivity, precision and freedom from analytical interferences.
A complete high-sensitivity plastic body nebulizer featuring Kalrez O-rings and Tantalum needle. The body is fabricated of inert plastic materials for maximum performance and corrosion resistance. It is designed for solutions containing aqua regia.
This assembly includes: impact bead, Qty.1 (97012-848), solvent resistant O-ring for nebulizer body, Qty.1 (97012-798), nebulizer body, Qty.1 (101676-644), spring, Qty.1 (97013-048), solvent resistant O-ring for capillary assembly, Qty.1 (97012-794), Pt/Ir capillary assembly with O-ring, Qty.1 (101684-716), locking ring, Qty.1 (101674-330), fine adjustment nut, Qty.1 (101674-332). It's compatible with the PerkinElmer AAnalyst® 200/400/700/800* and model 5100PC/5000/3300/3110/3100 AA instruments. Note: The AAnalyst® 800 requires end cap assembly.