Ilford 2000RT Machine Fixer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
83066-05
101381-668EA 147.02 USD
101381-668
Ilford 2000RT Machine Fixer, Electron Microscopy Sciences
Film Processing Supplies
The 2000RT is a non-hardening, liquid fixer concentrate specially designed for the rapid fixing of resin coated paper in processing machines complementing the 2000RT developer. It can be used at a wide range of fixer temperatures, 20 – 40°C (68 – 104°F). 2000RT Fixer must not be used with fix hardeners.