Ilford Hypam Fixer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
83062-50
101381-666EA 75.68 USD
101381-666
Ilford Hypam Fixer, Electron Microscopy Sciences
Film Processing Supplies
HYPAM is a non-hardening liquid rapid fixer that can be used with the fix hardening agents that are needed for some film processing applications. Without the addition of a hardener it is suitable for all black and white film and paper, manual and machine processing applications in the temperature range of 18 – 40°C (64 -104°F). HYPAM is the recommended fixer for all ILFORD scientific products and other X-ray films, plates and cineradiography films. If the addition of hardener is required, use ILFORD Hypam Hardener.