Ilford 2000RT Machine Developer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
83056-05
101381-658EA 184.83 USD
101381-658
Ilford 2000RT Machine Developer, Electron Microscopy Sciences
Film Processing Supplies
The 2000RT is a liquid concentrate developer/replenisher recommended for use with all types of black and white RC photo paper processors. It is easy to use as no starter solution is required. The recommended dilution for both the tank and replenisher solution is 1+4. It is a robust, economical, clean working developer with a long tank life. It can be used at a wide range of developing temperatures, 20 – 40°C (68 – 104°F). With most papers it gives a print image tone just slightly cool of neutral.