Ilford PQ Universal Dish/Tray Developer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
83052-50
101381-656EA 167.77 USD
101381-656
Ilford PQ Universal Dish/Tray Developer, Electron Microscopy Sciences
Film Processing Supplies
PQ UNIVERSAL is a liquid concentrate dimezones/hydroquinone developer which is suitable for all RC and FB black and white photographic papers. It is used at a dilution of 1+9, it is clean working and it has excellent keeping properties. It gives a slightly warm neutral image tone with most papers. In addition, PQ UNIVERSAL can be used to dish/tray process ILFORD as well as some other technical films. It is also suitable for dish/tray developing of general purpose sheet films when a fast working, high contrast developer is needed and a high degree of enlargement is not required. PQ UNIVERSAL developer is designed for use at ambient room temperatures, normally 20°C/68°F. We do not recommend its use for high temperature or machine processing applications.