Rheodyne™ 7725 Analytical Injection Valve, PerkinElmer
Supplier: PerkinElmer
N9306019 N9306017
97013-658EA 3054.93 USD
97013-658 10138-872
Rheodyne™ 7725 Analytical Injection Valve, PerkinElmer
Valves Injection Valves
Rheodyne™ 7725 and 7725i valves are versatile injectors and can use both partial-filling and complete-filling methods for loading the sample loop.
  • Improved access to fittings
  • Inject from 1μL to 5mL with high accuracy and precision