Thermal Conductivity Detector (TCD) 1/8" PPC Makeup Kit for PerkinElmer AutoSystem Series GCs, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N6120150
10138-730EA 4268.15 USD
10138-730
Thermal Conductivity Detector (TCD) 1/8" PPC Makeup Kit for PerkinElmer AutoSystem Series GCs, PerkinElmer
Chromatography Detectors
Includes parts required to add PPC controlled make-up gas to an existing Thermal Conductivity Detector (TCD).
Ordering information: Requires PPC capability in PerkinElmer AutoSystem XL GC. Includes tubing, tee-piece, PPC pneumatics module and PPC frit #4 (N6120155).