Quartz Reduction Tube, PerkinElmer
Supplier: PerkinElmer
N2411300
10138-428EA 229.43 USD
10138-428
Quartz Reduction Tube, PerkinElmer
Ovens Tube Furnaces
These quartz reduction tubes are composed of pure fused quartz which is 99% SiO₂. Fused quartz has been chosen because of its very high purity and low thermal expansion rate.
All combustion tubes are pressure tested to eliminate any tubes which might contain weak spots. In addition, strict specifications are used to ensure that the combustion tubes fit in an airtight connection when placed in the analyzer.