Tin Pans and Covers, PerkinElmer
Supplier: PerkinElmer
N2410320
10138-412EA 1864.37 USD
10138-412
Tin Pans and Covers, PerkinElmer
Trays/Dishes/Pots Laboratory Trays/Dishes
Pans with covers for volatile liquid or solid samples, including viscous materials.
Ordering information: Volatile sample sealer need to be ordered separately.