Non-Coded Intensitron Lamp Plug Adapter, PerkinElmer
Supplier: PerkinElmer
N3050197
10138-374EA 186.9 USD
10138-374
Non-Coded Intensitron Lamp Plug Adapter, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
Adapter allows non-coded Intensitron lamps (03036XXX series, 9-pin) and other 9-pin lamps to be used with PinAAcle and AAnalyst instruments (4-pin).
For use with legacy PerkinElmer Atomic Absorption (AA) instruments with Intensitron lamp connectors.