PolyPro-ST Nebulizer for PerkinElmer ICP-MS, PerkinElmer
Supplier: PerkinElmer
N0777220
10138-240EA 4520.22 USD
10138-240
PolyPro-ST Nebulizer for PerkinElmer ICP-MS, PerkinElmer
Atomizers
This PolyPro-ST nebulizer with detachable capillary is used with ELAN® and NexION® ICP-MS series instruments.
  • Polypropylene construction
  • Hydrofluoric acid resistant
  • 0 to 3 ml/min. sample flow rate with peristaltic pumping
  • Resistant to clogging and accidental breakage
The PolyPro-ST Nebulizer is an HF resistant alternative to the PFA nebulizer, but has the same high efficiency aerosol generation. It is a robust nebulizer that can self-aspirate with an exchangeable external sample uptake capillary, or it can be used with a peristaltic pump.