MiraMist® Nebulizer, PerkinElmer
Supplier: PerkinElmer
N0775330 N0777031
97013-704EA 2859.69 USD
97013-704 10138-194
MiraMist® Nebulizer, PerkinElmer
Atomizers
The Mira Mist® nebulizer handles most liquids, most flow ranges used in ICP and is one of the most rugged and longest life nebulizers due to its outer body being strong PEEK material. It fits regular spray chambers as a direct replacement for glass concentric nebulizers. Its features include standard pressure (45 psi), low sample flow and enhanced parallel path design.
  • The Mira Mist nebulizers are compatible with PerkinElmer Optima 4x00/5x00/7x00 and Avio 200/500 ICP-OES instruments
The PEEK Mira Mist nebulizer has an excellent mist and can handle a high level of particulates in the sample matrix.
The PTFE Mira Mist nebulizer is recommended for use with samples that contain high concentrations of Hydrofluroic and Sulfuric Acid and other solvents that attack PEEK.
Ordering information: The PEEK and PTFE models are sold separately.