O-Ring Free Glass Twister™ Spray Chamber with Helix for Optima 4300/5300/7300 V, PerkinElmer
Supplier: PerkinElmer
N0775356
10138-188EA 2283.53 USD
10138-188
O-Ring Free Glass Twister™ Spray Chamber with Helix for Optima 4300/5300/7300 V, PerkinElmer
Atomizers
This O-ring-free 50 ml borosilicate glass Twister™ spray chamber, with integrated Helix™, is used with Optima™ 4300 V, 5300 V, and 7300 V ICP-OES series instruments.
  • Material: borosilicate glass
  • Application uptake range 0.2 to 3.0 ml/min
  • Volume: 50 ml low RSD's due to highly accurate construction
The baffled, cyclonic Twister™ spray chamber has a central transfer tube, which acts as a secondary particle separator, helping separate larger aerosol particles from the sample. This reduces solvent load in the plasma without compromising detection limits. The position of the baffle is carefully controlled to ensure smooth draining and improved precision. The baffle is cut at a sharp angle so that a large cross-section of the spray chamber is sampled, ensuring representative sampling and reduced matrix effects. Although this spray chamber isn't hydrofluoric acid-resistant, precision is considered excellent and purity is considered good.
Produces smaller mean droplet size than standard spray chamber. Positive stop for reproducible nebulizer insertion convenient UniFit™ drain connector.