MicroMist™ Glass Nebulizer for ELAN® and NexION® Instruments, PerkinElmer
Supplier: PerkinElmer
N0775344
10138-172EA 2925.6 USD
10138-172
MicroMist™ Glass Nebulizer for ELAN® and NexION® Instruments, PerkinElmer
Atomizers
This glass MicroMist™ nebulizer includes connectors and sample tubing for use with ELAN® 5000/6x00/9000/DRCs and NexION® 300 and 350 ICP-MS series instruments. It can also be used with Optima™ 3x00 RL ICP-OES series instruments.
  • Glass concentric nebulizer offers high performance for limited sample volumes
  • Unit features a 0.05 ml/min. sample uptake rate
The MicroMist™ micro-uptake glass concentric nebulizer offers high performance for limited sample volumes. MicroMist™ nebulizers for ELAN® and NexION® 300/350 instruments are fitted with a UniFit connector with 1.3 mm O.D.×0.50 mm I.D.×700 mm long sample tubing.