Conikal™ U-Series Nebulizer 2 ml/min with EzyFit, PerkinElmer
Supplier: PerkinElmer
N0777487
10138-152EA 2026.8 USD
10138-152
Conikal™ U-Series Nebulizer 2 ml/min with EzyFit, PerkinElmer
Atomizers
This Conikal™ nebulizer, with 0.7 L/min argon flow rate and 2.0 ml/min uptake rate, is for use with Optima® 4x00/5x00/7x00 DV series, Optima® 8x00 and Avio® 200/500 ICP-OES series instruments. It is ideal for samples containing only moderate concentrations of dissolved salts, no particulates and no hydrofluoric acid.
Includes uniFit connector with Int. Ø 0.75×Ext. Ø 1.3 mm, Ext. Ø 700 mm long tubing (101675-616) and EzyLok connector kit (10139-382).