Optima Bench Cart, PerkinElmer
Supplier: PerkinElmer
N0771725
10138-120EA 10192.59 USD
10138-120
Optima Bench Cart, PerkinElmer
Carts
Optima bench for optical emission spectrometer.
  • Heavy-duty design
  • Locking casters
  • Cost-effective
  • Additional shelf for organizing consumables
This basic bench is a cost-effective option for storing Optima instruments. With an adjustable shelf designed for an autosampler, the autosampler can be configured to a position that is best suited. This bench also comes with a half storage shelf for organizing consumables and supplies.