Autosampler Probe with Filtered Tip, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N0771529
10138-118EA 1281.92 USD
10138-118
Autosampler Probe with Filtered Tip, PerkinElmer
Autosamplers
Stainless steel sample probe for autosamplers ASX-260, ASX-280, ASX-520, ASX-520HS, ASX-560, EXR-8, XLR-8, XLR-860, ASX-1400 and ASX-1600.
The stainless steel sample probe features a screen tip and is used in oils applications. The probe includes the capillary tube fitting, although does not include capillary tubing.