Glass Cyclonic Spray Chamber and Adapter Assembly for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Supplier: PerkinElmer
N0770605
10138-114EA 3532.4 USD
10138-114
Glass Cyclonic Spray Chamber and Adapter Assembly for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Atomizers
This glass cyclonic spray chamber and adaptor assembly is used with PerkinElmer Optima™ 2x00 DV, 4x00 DV, 5x00 DV, 7x00 DV and 8x00 ICP-OES series instruments.
Delivery information: Inclues glass standard cyclonic spray chamber (97012-938), cyclonic spray chamber mount kit (10134-360), cyclonic spray chamber mount nut, cyclonic spray chamber mount sleeve, cyclonic spray chamber mount ring, 20 mm M4 cap screw, 6 mm M3 cap screw, 3.2 mm M4 hex nut, round #8 thumb screw knob, polypropylene nipple barb fitting, Viton® O-ring 1.78 mm I.D., 3.175 mm I.D. Tygon® tubing (97012-046).