MEINHARD™ Nebulizers, 3 ml/min, PerkinElmer
Supplier: PerkinElmer
WE024371 N0681574
101687-282EA 1130.67 USD
101687-282 97012-894 10138-076
MEINHARD™ Nebulizers, 3 ml/min, PerkinElmer
Atomizers
The MEINHARD® concentric glass nebulizer reflects exacting standards in design and manufacturing. Its advantages include simplicity of design and operation, reproducible and self-aligning aerosol, and close fabrication control. For use with organics.