GemTip Cross-Flow Nebulizer Spares Kit for Optima 2x00/4x00/5x00/7x00, PerkinElmer
Supplier: PerkinElmer
N0680503
10138-070EA 1681.1 USD
10138-070
GemTip Cross-Flow Nebulizer Spares Kit for Optima 2x00/4x00/5x00/7x00, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This spares kit is designed for GemTip cross-flow nebulizers in Optima® 2x00/4x00/5x00/7x00 DV ICP-OES series instruments. It includes general replacement parts for the nebulizer.
Delivery information: This kit includes: O-Ring, 18W, 1³/₈ I.Ø., 1⁵/₈ OØ., Qty. 1 (09902033) Ferrule - Front ¹/₄, Qty. 1 (09903103) Ferrule - Back ¹/₄, Qty. 1 (09903104) Nut - Hex - 0.250 Tubing, Qty. 1 (09903199) Fitting Reducer ¹/₈ Tubing, ¹/₄ Port, Qty. 1 (09903471) Tubing (10 ft. ) - 0.023 I.Ø., Clear, Qty. 10 (09908265) Ferrule - Tubing ¹/₈ Tefzel, Qty. 1 (09920515) Ferrule - Tubing ¹/₁₆ Tefzel, Qty. 1 (09920518) Nut - Tubing Flanged 0.125 PEEK, Qty. 1 (09920545) O-Rings, 0.126 I.Ø., 0.040 W.D., Viton, Qty. 2 (09921045) Tubing, 0.032 IØ×0.016 Wall, PTFE, Qty. 2 ft (09985708) Argon Tubing, Qty. 2 ft (099857230) Tip Kit for Cross-Flow Nebulizer, Qty. 1 (N0690676) End Cap Cross-Flow Nebulizer, Qty. 1 (N8122239).