Adhesive Vinyl Labels 'Triple Rinsed', Labelmaster
Supplier: Label Master
TR333
101378-234PK 31.06 USD
101378-234
Adhesive Vinyl Labels 'Triple Rinsed', Labelmaster
Labels
These triple rinsed labels are used to identify containers that have been triple rinsed in accordance with EPA regulations in 40 CFR part 261.7.
  • Constructed of paper
  • Offer an economical alternative to vinyl
Color: White, blue and black