Labels 'Dry Ice', Labelmaster
Supplier: Label Master
HML11-DICE
101377-782PK 57.52 USD
101377-782
Labels 'Dry Ice', Labelmaster
Labels
Labelmaster’s paper Dry Ice Labels have UN1845 Dry Ice preprinted on them.
  • Meet the design and durability standards of 49 CFR, ICAO, and other international air regulations
  • Labels will not fade or fall off
  • Printed with light-fast inks for high durability
Labels feature blanks to fill in the dry ice quantity, the shipper's name, and the consignee. These Hazard Class 9 labels help maintain compliance with standards set forth by the DOT. Adhesives are formulated for use in various climates and environments. Hazard Class 9 paper labels have outstanding chemical and abrasion resistance, ensure strong adhesion to corrugated boxes, and help to keep users in regulatory compliance per the 49 CFR.